Anti-YAP1 Mouse Monoclonal Antibody [clone: 63.7]
Supplier: IMMUNOLOGIC
Certificates
About this item
Type: Primary
Antigen: YAP1
Clonality: Monoclonal
Clone: 63.7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Specifications
- Catalog No:
- ILM3920-C1
- ILM3920-C05
- ILM3920-C01
- Antigen symbol:
- YAP1
- YAP1
- YAP1
- Antigen name:
- Yes associated protein 1
- Yes associated protein 1
- Yes associated protein 1
- Antigen synonyms:
- YAP65
- YAP65
- YAP65
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- 63.7
- 63.7
- 63.7
- Reactivity:
- Human, Mouse
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 10413
- 10413
- 10413
- Isotype:
- IgG2a
- IgG2a
- IgG2a
- Western blot:
- Yes
- Yes
- Yes
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Form:
- Concentrated
- Concentrated
- Concentrated
- Molecular weight:
- 65 kDa
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C
- Tested applications:
- Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections
Specifications
Frequently Bought Together