Anti-WT1 Mouse Monoclonal Antibody [clone: 6F-H2]
Supplier: IMMUNOLOGIC
Certificates
About this item
Type: Primary
Antigen: WT1
Clonality: Monoclonal
Clone: 6F-H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- ILM3983-C01
- ILM3983-C1
- ILM3983-C05
- Antigen symbol:
- WT1
- WT1
- WT1
- Antigen name:
- Wilms tumor 1
- Wilms tumor 1
- Wilms tumor 1
- Antigen synonyms:
- WAGR|AWT1|NPHS4|WIT-2|GUD
- WAGR|AWT1|NPHS4|WIT-2|GUD
- WAGR|AWT1|NPHS4|WIT-2|GUD
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- 6F-H2
- 6F-H2
- 6F-H2
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 4790
- 4790
- 4790
- Isotype:
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Yes
- Immunogen:
- Recombinant protein corresponding to residues 1-181 of human WT1
- Form:
- Concentrated
- Concentrated
- Concentrated
- Molecular weight:
- 47-55 kDa
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C
- Tested applications:
- Immunohistochemical Staining of Formalin-Fixed Tissue Sections
Specifications
Frequently Bought Together