Anti-DNTT Rabbit Polyclonal Antibody
Supplier: IMMUNOLOGIC
Certificates
About this item
Type: Primary
Antigen: DNTT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Calf, Rat, Mouse, Guinea pig, Cat, Chicken
Specifications
- Catalog No:
- ILP0049-C01
- ILP0049-C05
- ILP0049-C1
- Antigen symbol:
- DNTT
- DNTT
- DNTT
- Antigen name:
- DNA nucleotidylexotransferase
- DNA nucleotidylexotransferase
- DNA nucleotidylexotransferase
- Antigen synonyms:
- TDT
- TDT
- TDT
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Reactivity:
- Chicken, Calf, Human, Guinea pig, Rat, Mouse, Cat
- Chicken, Calf, Human, Guinea pig, Rat, Mouse, Cat
- Chicken, Calf, Human, Guinea pig, Rat, Mouse, Cat
- Host:
- Rabbit
- Rabbit
- Rabbit
- Gene ID:
- 1791
- 1791
- 1791
- Isotype:
- IgG
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Form:
- Concentrated
- Concentrated
- Concentrated
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C
Specifications
Frequently Bought Together