Anti-SOX10 Mouse Monoclonal Antibody [clone: SOX10/1074]
Supplier: IMMUNOLOGIC
Certificates
About this item
Type: Primary
Antigen: SOX10
Clonality: Monoclonal
Clone: SOX10/1074
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Specifications
- Catalog No:
- ILM6663-C1
- VWRKILM6663-C05
- Antigen symbol:
- SOX10
- SOX10
- Antigen name:
- SRY-box 10
- SRY-box 10
- Antigen synonyms:
- WS4|DOM|WS2E
- WS4|DOM|WS2E
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- SOX10/1074
- SOX10/1074
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 6663
- 6663
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Form:
- Concentrated
- Concentrated
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
Specifications
Frequently Bought Together