Anti-TP53 Mouse Monoclonal Antibody [clone: BP53-12 ]
Supplier: IMMUNOLOGIC
Certificates
About this item
Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: BP53-12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey, Dog, Hamster, Chicken
Specifications
- Catalog No:
- ILM7157-C05
- ILM7157-C01
- ILM7157-C1
- Antigen symbol:
- TP53
- TP53
- TP53
- Antigen name:
- Tumor protein p53
- Tumor protein p53
- Tumor protein p53
- Antigen synonyms:
- LFS1|p53
- LFS1|p53
- LFS1|p53
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- BP53-12
- BP53-12
- BP53-12
- Reactivity:
- Hamster, Monkey, Chicken, Human, Dog
- Hamster, Monkey, Chicken, Human, Dog
- Hamster, Monkey, Chicken, Human, Dog
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 7157
- 7157
- 7157
- Isotype:
- IgG2a
- IgG2a
- IgG2a
- Western blot:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Yes
- Form:
- Concentrated
- Concentrated
- Concentrated
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C
Specifications
Frequently Bought Together