Anti-POU5F1 Mouse Monoclonal Antibody [clone: C-10]
Supplier: IMMUNOLOGIC
Certificates
About this item
Type: Primary
Antigen: POU5F1
Clonality: Monoclonal
Clone: C-10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Specifications
- Catalog No:
- ILM527919-C1
- ILM527919-C05
- ILM527919-C01
- Antigen symbol:
- POU5F1
- POU5F1
- POU5F1
- Antigen name:
- POU class 5 homeobox 1
- POU class 5 homeobox 1
- POU class 5 homeobox 1
- Antigen synonyms:
- MGC22487|Oct4|OCT3|OTF3
- MGC22487|Oct4|OCT3|OTF3
- MGC22487|Oct4|OCT3|OTF3
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- C-10
- C-10
- C-10
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 5460
- 5460
- 5460
- Isotype:
- IgG2b
- IgG2b
- IgG2b
- Western blot:
- Yes
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- ImmunoFluorescence:
- Yes
- Gel shift:
- Yes
- ChIP:
- Yes
- Epitope:
- C-terminus
- Form:
- Concentrated
- Concentrated
- Concentrated
- Molecular weight:
- 52 kDa (Oct-3/4A isoform); 45 kDa (Oct-3/4B isoform)
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C
- Tested applications:
- Immunohistochemical Staining of Paraffin Embedded Tissue Sections
Specifications
Frequently Bought Together