Anti-MDM2 Mouse Monoclonal Antibody [clone: SMP14]
Type: Primary
Antigen: MDM2
Clonality: Monoclonal
Clone: SMP14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
- Catalog No:
- ILM5324-C1
- ILM5324-C05
- ILM5324-C01
- Antigen symbol:
- MDM2
- MDM2
- MDM2
- Antigen name:
- MDM2 proto-oncogene
- MDM2 proto-oncogene
- MDM2 proto-oncogene
- Antigen synonyms:
- HDM2|MGC5370
- HDM2|MGC5370
- HDM2|MGC5370
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- SMP14
- SMP14
- SMP14
- Reactivity:
- Human, Rat, Mouse
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 4193
- 4193
- 4193
- Isotype:
- IgG1
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Yes
- Immunogen:
- Human MDM2 aa. 154-167
- Form:
- Concentrated
- Concentrated
- Concentrated
- Molecular weight:
- 95 kDa
- Purification:
- Affinity chromatography
- Storage buffer:
- Aqueous buffered solution containing ?0,09% sodium azide
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C
- Tested applications:
- Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections, Bioimaging
Frequently Bought Together