Anti-Cytokeratin Mouse Monoclonal Antibody [clone: AE1+AE3]
Type: Primary
Antigen: Cytokeratin
Clonality: Monoclonal
Clone: AE1+AE3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey, Cow, Dog, Rabbit, Mouse, Rat, Chicken
- Catalog No:
- ILM3133-C1
- ILM3133-C05
- ILM3133-C01
- Antigen symbol:
- KRT1/KRT3/KRT4/KRT5/KRT6/KRT10/KRT14/KRT15/KRT16/KRT19
- KRT1/KRT3/KRT4/KRT5/KRT6/KRT10/KRT14/KRT15/KRT16/KRT19
- KRT1/KRT3/KRT4/KRT5/KRT6/KRT10/KRT14/KRT15/KRT16/KRT19
- Antigen name:
- Keratin 1/Keratin 3/Keratin 4/Keratin 5/Keratin 6/Keratin 10/Keratin 14/Keratin 15/Keratin 16/Keratin 19
- Keratin 1/Keratin 3/Keratin 4/Keratin 5/Keratin 6/Keratin 10/Keratin 14/Keratin 15/Keratin 16/Keratin 19
- Keratin 1/Keratin 3/Keratin 4/Keratin 5/Keratin 6/Keratin 10/Keratin 14/Keratin 15/Keratin 16/Keratin 19
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- AE1+AE3
- AE1+AE3
- AE1+AE3
- Reactivity:
- Rabbit, Monkey, Chicken, Human, Bovine, Dog, Rat, Mouse
- Rabbit, Monkey, Chicken, Human, Bovine, Dog, Rat, Mouse
- Rabbit, Monkey, Chicken, Human, Bovine, Dog, Rat, Mouse
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa
- ELISA:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Yes
- Form:
- Concentrated
- Concentrated
- Concentrated
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C
Frequently Bought Together