Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-KRT20 Rabbit Monoclonal Antibody [clone: E19-1]
 
Anti-KRT20 Rabbit Monoclonal Antibody [clone: E19-1]

 

  • Catalog No:
  • ILM2133-C1
  • ILM2133-C05
  • ILM2133-C01
  • Antigen symbol:
  • KRT20
  • KRT20
  • KRT20
  • Antigen name:
  • Keratin 20
  • Keratin 20
  • Keratin 20
  • Antigen synonyms:
  • K20|CK20|MGC35423
  • K20|CK20|MGC35423
  • K20|CK20|MGC35423
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • E19-1
  • E19-1
  • E19-1
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Gene ID:
  • 54474
  • 54474
  • 54474
  • Isotype:
  • IgG
  • IgG
  • IgG
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • Epitope:
  • C-terminal
  • Immunogen:
  • Peptide derived from C-terminal region of human cytokeratin 20. Antibody recognizes the epitope
    between Val408 - Ile424
  • Form:
  • Concentrated
  • Concentrated
  • Concentrated
  • Storage temperature:
  • 2°C - 8°C
  • 2°C - 8°C
  • 2°C - 8°C

 

Frequently Bought Together