Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-CEACAM5 Mouse Polyclonal Antibody [clone: COL-1]
 
Anti-CEACAM5 Mouse Polyclonal Antibody [clone: COL-1]

 

  • Catalog No:
  • ILM83111-C1
  • ILM83111-C05
  • ILM83111-C01
  • Antigen symbol:
  • CEACAM5
  • CEACAM5
  • CEACAM5
  • Antigen name:
  • Carcinoembryonic antigen related 
  • Carcinoembryonic antigen related 
  • Carcinoembryonic antigen related 
  • Antigen synonyms:
  • CEA|CD66e
  • CEA|CD66e
  • CEA|CD66e
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • COL-1
  • COL-1
  • COL-1
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 1048
  • 1048
  • 1048
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • IgG2a kappa
  • Western blot:
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Form:
  • Concentrated
  • Concentrated
  • Concentrated
  • Molecular weight:
  • 80-200 kDa
  • Storage temperature:
  • 2°C - 8°C
  • 2°C - 8°C
  • 2°C - 8°C
  • Tested applications:
  • Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections

 

Frequently Bought Together