Anti-B3GAT1 Mouse Monoclonal Antibody [clone: NK-1]
Supplier: IMMUNOLOGIC
Certificates
About this item
Type: Primary
Antigen: B3GAT1
Clonality: Monoclonal
Clone: NK-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human
Specifications
- Catalog No:
- ILM1021-C1
- ILM1021-C05
- ILM1021-C01
- Antigen symbol:
- B3GAT1
- B3GAT1
- B3GAT1
- Antigen name:
- Beta-1,3-glucuronyltransferase 1
- Beta-1,3-glucuronyltransferase 1
- Beta-1,3-glucuronyltransferase 1
- Antigen synonyms:
- LEU7|CD57|HNK-1|NK-1|GlcAT-P
- LEU7|CD57|HNK-1|NK-1|GlcAT-P
- LEU7|CD57|HNK-1|NK-1|GlcAT-P
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- NK-1
- NK-1
- NK-1
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 27087
- 27087
- 27087
- Isotype:
- IgM kappa
- IgM kappa
- IgM kappa
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Yes
- Form:
- Concentrated
- Concentrated
- Concentrated
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C
Specifications
Frequently Bought Together