Order Entry
Netherlands
ContactUsLinkComponent
Anti-CALD1 Mouse Monoclonal Antibody [clone: H-CALD]
Anti-CALD1 Mouse Monoclonal Antibody [clone: H-CALD]
Supplier:  IMMUNOLOGIC
Certificates
Anti-CALD1 Mouse Monoclonal Antibody [clone: H-CALD]
Supplier:  IMMUNOLOGIC

Specifications

  • Catalog No:
  • ILM8029-C1
  • ILM8029-C05
  • ILM8029-C01
  • Antigen symbol:
  • CALD1
  • CALD1
  • CALD1
  • Antigen name:
  • Caldesmon1
  • Caldesmon1
  • Caldesmon1
  • Antigen synonyms:
  • L-CAD|CDM|H-CAD
  • L-CAD|CDM|H-CAD
  • L-CAD|CDM|H-CAD
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • H-CALD
  • H-CALD
  • H-CALD
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 800
  • 800
  • 800
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Form:
  • Concentrated
  • Concentrated
  • Concentrated
  • Molecular weight:
  • 150 kDa
  • Storage temperature:
  • 2°C - 8°C
  • 2°C - 8°C
  • 2°C - 8°C

Specifications

Related Information

Frequently Bought Together