Order Entry
Netherlands
ContactUsLinkComponent
Anti-BCA-225 Mouse Monoclonal Antibody [clone: CU-18]
Anti-BCA-225 Mouse Monoclonal Antibody [clone: CU-18]
Supplier:  IMMUNOLOGIC
Certificates
Anti-BCA-225 Mouse Monoclonal Antibody [clone: CU-18]
Supplier:  IMMUNOLOGIC

Specifications

  • Catalog No:
  • ILM2122-C1
  • ILM2122-C05
  • ILM2122-C01
  • Antigen symbol:
  • BCA-225
  • BCA-225
  • BCA-225
  • Antigen name:
  • BCA-225 molecule
  • BCA-225 molecule
  • BCA-225 molecule
  • Antigen synonyms:
  • BRST1
  • BRST1
  • BRST1
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • CU-18
  • CU-18
  • CU-18
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Isotype:
  • IgG kappa
  • IgG kappa
  • IgG kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • A synthetic peptide
  • Form:
  • Concentrated
  • Concentrated
  • Concentrated
  • Storage temperature:
  • 2°C - 8°C
  • 2°C - 8°C
  • 2°C - 8°C
  • Tested applications:
  • Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections

Specifications

Related Information

Frequently Bought Together