Anti-BAP1 Mouse Monoclonal Antibody [clone: C-4]
Supplier: IMMUNOLOGIC
Certificates
About this item
Type: Primary
Antigen: BAP1
Clonality: Monoclonal
Clone: C-4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- ILM3919-C1
- ILM3919-C05
- ILM3919-C01
- Antigen symbol:
- BAP1
- BAP1
- BAP1
- Antigen name:
- BRCA1 associated protein 1
- BRCA1 associated protein 1
- BRCA1 associated protein 1
- Antigen synonyms:
- KIAA0272|hucep-6|UCHL2
- KIAA0272|hucep-6|UCHL2
- KIAA0272|hucep-6|UCHL2
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- C-4
- C-4
- C-4
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 8314
- 8314
- 8314
- Isotype:
- IgG1
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Form:
- Concentrated
- Concentrated
- Concentrated
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C
Specifications
Frequently Bought Together