Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standard, quizalofop-P-tefuryl 10 µg/ml in acetonitrile, Dr. Ehrenstorfer
Organic reference standard, quizalofop-P-tefuryl 10 µg/ml in acetonitrile, Dr. Ehrenstorfer
Catalog # EHERL16741000AL
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, quizalofop-P-tefuryl 10 µg/ml in acetonitrile, Dr. Ehrenstorfer
Catalog # EHERL16741000AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Standards:
    Reference standard
  • CAS No.:
    200509-41-7
  • Description:
    Quizalofop-P-tefuryl 10 µg/ml in acetonitrile
  • Formula:
    C₂₂H₂₁ClN₂O₅
  • Description:
    Quizalofop-P-tefuryl

Specifications

About this item

Certifications: Accredited to ISO/IEC 17025.