

Methenamine ≥99.0% for synthesis, Sigma-Aldrich®
: Merck
:100-97-0
:
:C₆H₁₂N₄
:MFCD00006895
:13,05994
:280 °C
:1,33 g/cm³ (20 °C)
:250 °C
:202-905-8
:1328
:4.1,III
:M-Clarity™ quality level = MQ200
:140.19 g/mol
:GHS02
:GHS07
:Kamertemperatuur
- Assay (HClO₄):≥99.0%
- Identity (IR):Passes Test
- Bulk Density:600 kg/m3
- Vapor Pressure:(20 °C)
- Flash Point:250 °C
- Density:1.33 g/cm3 (20 °C)
- pH:7.0 - 9.0 (100 g/l, H2O, 20 °C)
- Ignition Point:390 °C
- Hazard Class:4.1
- Hazard Class Description:Flammable Solid
Frequently Bought Together