Rat Recombinant MCP-3 (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Monocyte chemotactic protein 3 (MCP-3), also called CCL7, is produced by macrophages and tumor cell lines. MCP-3 signals through the G protein-coupled receptors CCR1, CCR2, and CCR3. MCP-3 chemoattracts monocytes and regulates macrophage function during inflammation and metastasis.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q9QXY8
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:MARC|CCL7
- Protein/peptide name:MCP-3
- Purity:≥95%
- Molecular weight:8,5 kDa
- Sequence:QPDGTNSSTC CYVKKQKIPK RNLKSYRKIT SSRCPWEAVI FKTKKGMEVC AEAHQKWVEE AIAYLDMKTS TPKP
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together