2-Bromo-3-hydroxypyridine ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 6602-32-0 Molecular Formula: C5H4BrNO Formula Weight: 174.00 Melting Point: 182-186° Beilstein Reference: 109829
Synonyms: 2-Bromo-3-pyridinol
- CAS Number:6602-32-0
- Formula:C₅H₄BrNO
- MDL Number:MFCD00006220
- Molecular Weight:174 g/mol
- Boiling Pt:232 °C (1013 hPa)
- Melting Pt:182…186 °C
- Storage Temperature:Ambient
- EINECS:229-547-5
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 6602-32-0 Molecular Formula: C5H4BrNO Formula Weight: 174.00 Melting Point: 182-186° Beilstein Reference: 109829
Synonyms: 2-Bromo-3-pyridinol
- CAS Number:6602-32-0
- Formula:C₅H₄BrNO
- MDL Number:MFCD00006220
- Molecular Weight:174 g/mol
- Boiling Pt:232 °C (1013 hPa)
- Melting Pt:182…186 °C
- Storage Temperature:Ambient
- EINECS:229-547-5



