

1-Methyl-1H-pyrazole ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:930-36-9
:
Formula:C₄H₆N₂
MDL Number:MFCD00144943
:82.11 g/mol
Boiling Pt:127°C
Density:0,993 g/cm³ (20 °C)
Flash Pt:36°C
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together