

1-Methyl-1H-pyrazole 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:930-36-9
:
Formula:C₄H₆N₂
MDL Number:MFCD00144943
:82.11 g/mol
Boiling Pt:127 °C (760 mmHg)
Density:0.993 g/cm³
Flash Pt:35 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4760 to 1.4780 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together