Hypoxanthine
Glass bottle for solids, 25 g
Catalog # SANTSC-29068
Marketsource
Supplier: Santa Cruz Biotechnology
Hypoxanthine
Catalog # SANTSC-29068
Supplier: Santa Cruz Biotechnology
Marketsource
CAS Number: 68-94-0
Jump to:
Product Details & Documents
Synonyms: 1H-Purin-6-one, 1,9-Dihydro-6H-purin-6-one, Purin-6(1H)-one, 6-Purinone
- CAS Number:68-94-0
- Formula:C₅H₄N₄O
- MDL Number:MFCD00005725
- Molecular Weight:136.11 g/mol
- Merck Index:13,04895
- Melting Pt:> 310 °C
- EINECS:200-697-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



