Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Hypoxanthine, Millipore®

Hypoxanthine, Millipore®

Glass bottle for solids, 25 g
Catalog # 4010CBC-25GM
Hypoxanthine, Millipore®
Catalog # 4010CBC-25GM
CAS Number:  68-94-0
Hypoxanthine, Millipore®

Glass bottle for solids, 25 g

CAS Number:  68-94-0
Price
108,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1H-Purin-6-one, 1,9-Dihydro-6H-purin-6-one, Purin-6(1H)-one, 6-Purinone
  • CAS Number:68-94-0
  • Formula:C₅H₄N₄O
  • MDL Number:MFCD00005725
  • Molecular Weight:136.11 g/mol
  • Merck Index:13,04895
  • Melting Pt:> 310 °C
  • EINECS:200-697-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications