Hypoxanthine ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 68-94-0 Molecular Formula: C5H4N4O Formula Weight: 136.11 Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 5811 Merck Reference: 14,4869 MDL No.: MFCD00005725
Synonyms: 1H-Purin-6-one, 1,9-Dihydro-6H-purin-6-one, Purin-6(1H)-one, 6-Purinone
- CAS Number:68-94-0
- Formula:C₅H₄N₄O
- MDL Number:MFCD00005725
- Molecular Weight:136.11 g/mol
- Merck Index:14,4869
- Melting Pt:>300 °C
- EINECS:200-697-3
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 68-94-0 Molecular Formula: C5H4N4O Formula Weight: 136.11 Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 5811 Merck Reference: 14,4869 MDL No.: MFCD00005725
Synonyms: 1H-Purin-6-one, 1,9-Dihydro-6H-purin-6-one, Purin-6(1H)-one, 6-Purinone
- CAS Number:68-94-0
- Formula:C₅H₄N₄O
- MDL Number:MFCD00005725
- Molecular Weight:136.11 g/mol
- Merck Index:14,4869
- Melting Pt:>300 °C
- EINECS:200-697-3
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



