
15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:33100-27-5
:
Formula:C₁₀H₂₀O₅
MDL Number:MFCD00005110
Boiling Pt:150…155 °C (0.5 mmHg)
Melting Pt:–20 °C
Density:1.110
Flash Pt:>110 °C (230 °F)
:220.27 g/mol
EINECS:251-379-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together