Fibroblast growth factor 4 (FGF-4) is a secreted growth factor that is predominantly expressed during bone morphogenesis and embryonic limb development. FGF-4 is an important growth regulator for stem cells, fibroblasts, and endothelial cells. FGF-4 contains a single N-linked glycosylation signal. In-vitro studies suggest that unglycosylated FGF-4 is cleaved into 13 kDa and 15 kDa truncated proteins that have greater biological activity than the wild type 19 kDa FGF-4 protein. Human FGF-4 shares high homology and is cross-reactive with mouse FGF-4.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity assay: NR6R-3T3 Proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2,0×10⁵ units/mg
- Gene ID:P08620
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Transforming protein KS3|HST-1|HBGF-4
- Protein/peptide name:FGF-4
- Purity:≥95%
- Molecular weight:19,4 kDa
- Sequence:MAPTAPNGTL EAELERRWES LVALSLARLP VAAQPKEAAV QSGAGDYLLG IKRLRRLYCN VGIGFHLQAL PDGRIGGAHA DTRDSLLELS PVERGVVSIF GVASRFFVAM SSKGKLYGSP FFTDECTFKE ILLPNNYNAY ESYKYPGMFI ALSKNGKTKK GNRVSPTMKV THFLPRL
- Endotoxin level:Low
- Formulation:20 mM sodium phosphate, 0,5 M sodium chloride, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay