Human recombinant ENA 78/CXCL5 (from E. coli)
Epithelial-derived neutrophil-activating peptide 78 (ENA 78), also known as CXCL5, is a chemokine that recruits neutrophils, promotes angiogenesis.
- Produced from genetically modified E. coli
- Stimulates connective tissue remodelling
- Monomeric protein, contains 74 amino acids
ENA 78 production is stimulated by interleukin 1 (IL-1) or tumor necrosis factor alpha (TNFα), and signals through the chemokine receptor CXCR2.
ENA 78, 5-78 aa is one of three naturally occurring ENA 78 variants in which the N-terminus has been truncated.
: A Certificate of Analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
: This product is for research use only and is not for use in humans.
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Form:Lyophilised
- Gene ID:P42830
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CXCL5|Epithelial-derived neutrophil-activating protein 78
- Protein/peptide name:ENA 78 / CXCL5 (5-78 aa)
- Purity:≥95%
- Molecular weight:8,1 kDa
- Sequence:AAVLRELRCV CLQTTQGVHP KMISNLQVFA IGPQCSKVEV VASLKNGKEI CLDPEAPFLK KVIQKILDGG NKEN
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Frequently Bought Together