Tantalum(V) oxide ≥99.993% (metals basis excluding Nb), Puratronic® (max. 0.005% Nb)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1314-61-0
:
Formula:O₅Ta₂
MDL Number:MFCD00011254
Merck Index:14,9056
Melting Pt:1800 °C
Density:8.2
:441.89 g/mol
Storage Temperature:Ambient
EINECS:215-238-2
Frequently Bought Together