Organic reference standard, Florfenicol amine, VETRANAL™
32492-10MG
: Merck
:
- Pk:10 mg
- Standards:Reference standard
- CAS No.:76639-93-5
- Description:Florfenicol amine
- Formula:C₁₀H₁₄FNO₃S
- Description:Florfenicol amine
- Suitable for HPLC and gas chromatography (GC)