Yttrium(III) fluoride ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 13709-49-4 Molecular Formula: F3Y Formula Weight: 145.90 Physical Form: -325 Mesh Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 1387° UN No.: 3288 MDL No.: MFCD00049714
Synonyms: Yttrium trifluoride, Yttrium(III) fluoride
- CAS Number:13709-49-4
- Formula:F₃Y
- MDL Number:MFCD00049714
- Molecular Weight:145.9 g/mol
- Boiling Pt:2227 °C (1013 hPa)
- Melting Pt:1387 °C
- Density:4.01 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:237-257-5
- UN:3288
- ADR:6.1,III
- :Density: 4.01 g/cm³ (20 °C)
2 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 13709-49-4 Molecular Formula: F3Y Formula Weight: 145.90 Physical Form: -325 Mesh Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 1387° UN No.: 3288 MDL No.: MFCD00049714
Synonyms: Yttrium trifluoride, Yttrium(III) fluoride
- CAS Number:13709-49-4
- Formula:F₃Y
- MDL Number:MFCD00049714
- Molecular Weight:145.9 g/mol
- Boiling Pt:2227 °C (1013 hPa)
- Melting Pt:1387 °C
- Density:4.01 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:237-257-5
- UN:3288
- ADR:6.1,III
- :Density: 4.01 g/cm³ (20 °C)



