Naphthalene
Glass bottle for solids, 500 mg
Catalog # PROOLGC2603
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
- CAS Number:91-20-3
- Formula:C₁₀H₈
- MDL Number:MFCD00001742
- Molecular Weight:128.17 g/mol
- Merck Index:12,06457
- Boiling Pt:218 °C (1013 hPa)
- Melting Pt:79…82 °C
- Density:1.145 g/cm³ (20 °C)
- Flash Pt:79 °C
- Storage Temperature:Ambient
- EINECS:202-049-5
- UN:1334
- ADR:4.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg



