
1-Naphthyl isocyanate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:86-84-0
:
Formula:C₁₁H₇NO
MDL Number:MFCD00003881
Merck Index:12,06497
Boiling Pt:269…270 °C
Melting Pt:4 °C
Density:1.18 g/cm³
Flash Pt:135 °C
UN:2206
ADR:6.1,III
:169.18 g/mol
EINECS:201-703-7
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6324 to 1.6344 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together