

Dilanthanum trisulphide ≥99% (REO, rare earth oxide basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12031-49-1
:
Formula:La₂S₃
MDL Number:MFCD00016168
:374.01 g/mol
Melting Pt:2100 °C in vac.
Density:4.911
Storage Temperature:Ambient
EINECS:234-752-8
UN:3134
ADR:4.3,III
:GHS02
:GHS07
Related Information
Frequently Bought Together