Neodymium(III) oxide ≥99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1313-97-9
:
Formula:Nd₂O₃
MDL Number:MFCD00011134
:336.48 g/mol
Merck Index:14,6450
Boiling Pt:3760 °C (1013 hPa)
Melting Pt:2272 °C
Density:7.24
Storage Temperature:Ambient
EINECS:215-214-1
Frequently Bought Together