
Europium(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13765-25-8
:
Formula:EuF₃
MDL Number:MFCD00010998
:208.96 g/mol
Boiling Pt:>2280 °C
Melting Pt:1390 °C
Storage Temperature:Ambient
EINECS:237-368-9
:GHS07
Related Information
Frequently Bought Together