
Dibutyl disulphide for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:629-45-8
:
Formula:C₈H₁₈S₂
MDL Number:MFCD00009467
:178.36 g/mol
Boiling Pt:229 °C (1013 hPa)
Melting Pt:–94 °C
Density:0.9371 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:211-091-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:-94 °C
- Molar Mass:178.35 g/mol
- Boiling Point:229 - 233 °C
- Flash Point:93 °C
- Refractive Index:1.492 (20 °C, 589 nm)
- Explosion Limit:0.8 %(V)
- Density:0.94 g/cm3 (20 °C)
- Synonyms:Butyl disulfide
- Hill Formula:C8H18S2
- Chemical Formula:CH3(CH2)3SS(CH2)3CH3
Specification Test Results
Frequently Bought Together