
Ammonium hydrogen carbonate
Supplier: Roth Carl
Certificates
About this item
CAS Number:1066-33-7
:
Formula:NH₄HCO₃
MDL Number:MFCD00012138
:79.06 g/mol
Merck Index:13,00497
Melting Pt:35 °C
Density:1,57 g/cm³ (20 °C)
EINECS:213-911-5
:GHS07
Frequently Bought Together