Ammonium chloride
Marketsource
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Ammonium muriate, Salmiac
- CAS Number:12125-02-9
- Formula:NH₄Cl
- MDL Number:MFCD00011420
- Molecular Weight:53.49 g/mol
- Merck Index:13,00510
- Melting Pt:335 °C
- Density:1.5256 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:235-186-4
3 Options Available Below
Product Details & Documents
Synonyms: Ammonium muriate, Salmiac
- CAS Number:12125-02-9
- Formula:NH₄Cl
- MDL Number:MFCD00011420
- Molecular Weight:53.49 g/mol
- Merck Index:13,00510
- Melting Pt:335 °C
- Density:1.5256 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:235-186-4
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



