Order Entry
Netherlands
ContactUsLinkComponent
Tetrabutylammonium fluoride ∼1 mol/L in THF for catalyst of silylation, for cleavage of silyl ether
Flammable
Corrosive
Irritant
Health
Tetrabutylammonium fluoride ∼1 mol/L in THF for catalyst of silylation, for cleavage of silyl ether
Supplier:  TCI
Certificates
undefined
Flammable
Corrosive
Irritant
Health
Tetrabutylammonium fluoride ∼1 mol/L in THF for catalyst of silylation, for cleavage of silyl ether
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Yellow to Green clear liquid
  • Concentration (Nonaqueous Titration):
    1.0 to 1.1 mol/L
  • Water:
    max. 7.0 %
  • TBHF-HF (neutralization titration):
    max. 2.0 %

Specification Test Results

Related Information

Frequently Bought Together