
(1-Tetradecyl)triphenylphosphonium bromide ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25791-20-2
:
Formula:H₃C(CH₂)₁₃P(C₆H₅)₃Br
MDL Number:MFCD00051857
:539.58 g/mol
Melting Pt:87…89 °C
EINECS:247-267-1
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together