Triethyl phosphate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:78-40-0
:
Formula:C₆H₁₅O₄P
MDL Number:MFCD00009077
:182.16 g/mol
Merck Index:12,09806
Boiling Pt:215 °C (1013 hPa)
Melting Pt:–57 °C
Density:1.07 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:116 °C
EINECS:201-114-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:500 g/l (25 °C) (slow decomposition)
- Melting Point:-57 °C
- Molar Mass:182.16 g/mol
- Boiling Point:215 °C (1013 hPa)
- Vapor Pressure:0.1 hPa (20 °C)
- Flash Point:130 °C
- Refractive Index:1.4053 (20 °C, 589 nm)
- Explosion Limit:1.2 - 10 %(V)
- Density:1.069 g/cm3 (20 °C)
- pH:7 (H2O, 20 °C)
- Ignition Point:480 °C
Specification Test Results
Related Information
Certificates
Search by Lot Numbermsds_821141
SDS
ProductPage_821141 ProductPage_821141
Product specification
Frequently Bought Together