Triisopropylchlorosilane
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:13154-24-0
:
Formula:C₉H₂₁ClSi
MDL Number:MFCD00009656
:192.8 g/mol
Boiling Pt:198 °C (1013 hPa)
Density:0.9008 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2987
ADR:8,II
:GHS05
Frequently Bought Together