
Octyltriethoxysilane 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2943-75-1
:
Formula:C₁₄H₃₂O₃Si
MDL Number:MFCD00039883
:276.49 g/mol
Boiling Pt:265 °C (1013 hPa)
Melting Pt:–40 °C
Density:0.88 g/cm³
Flash Pt:120 °C
EINECS:220-941-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4160 to 1.4180 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together