
Octadecyltrimethoxysilane 90% tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3069-42-9
:
Formula:C₂₁H₄₆O₃Si
MDL Number:MFCD00043060
:374.68 g/mol
Boiling Pt:170 °C (.1 mmHg)
Melting Pt:13…17 °C
Density:0.88 g/cm³
Flash Pt:139 °C
EINECS:221-339-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear to slightly hazy colorless to light yellow Liquid (May develop some turbidity or precipitate. May partially crystallize)
- Infrared spectrum:Conforms
- GC:≥88 % Total octadecyltrimethoxysilanes
Specification Test Results
Frequently Bought Together