
N-(3-(Trimethoxysilyl)propyl)ethylenediamine tech. 90%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1760-24-3
:
Formula:H₂NCH₂CH₂NHCH₂CH₂CH₂Si(OCH₃)₃
MDL Number:MFCD00008173
:222.36 g/mol
Boiling Pt:261…263 °C
Melting Pt:∼ 0 °C
Density:1.010
Flash Pt:136 °C (276 °F)
Storage Temperature:Ambient
EINECS:217-164-6
UN:2735
ADR:8,III
:GHS05
Related Information
Frequently Bought Together