(3-Chloropropyl)trimethoxysilane for synthesis, Sigma-Aldrich®
Catalog # 8.18816.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 ml
- Seq:2590
Specifications
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Melting Point:-50 °C
- Molar Mass:198.72 g/mol
- Boiling Point:195 - 197 °C (1013 hPa)
- Vapor Pressure:- 5 hPa (20 °C)
- Flash Point:78 °C
- Refractive Index:1.4242 (25 °C, 589 nm)
- Explosion Limit:5.5 - 44 %(V)
- Density:1.082 g/cm3 (20 °C)
- Ignition Point:220 °C
- Synonyms:CPTMO
- Hill Formula:C6H15ClO3Si
- Chemical Formula:Cl(CH2)3Si(OCH3)3
Specification Test Results
About this item
:
:198.72 g/mol
Boiling Pt:195 °C (1013 hPa)
Melting Pt:–50 °C
Density:1.082 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:219-787-9
CAS Number:2530-87-2
Formula:ClH₂CCH₂CH₂Si(OCH₃)₃
MDL Number:MFCD00000997