
Triisopropyl borate for synthesis, Sigma-Aldrich®
: Merck
:5419-55-6
:
:C₉H₂₁BO₃
:MFCD00008872
:188.07 g/mol
:139 °C (1013 hPa)
:–59 °C
:0.82 g/cm³ (20 °C)
:226-529-9
:3,II
:M-Clarity™ quality level = MQ200
:2616
:GHS02
:Ambient
- Melting Point:-59 °C
- Molar Mass:188.07 g/mol
- Boiling Point:139 - 141 °C (1013 hPa)
- Vapor Pressure:75 hPa (100 °C)
- Flash Point:25.5 °C
- Density:0.82 g/cm3 (20 °C)
- Hazard Class:3
- Hazard Class Description:Flammable liquid
Frequently Bought Together