

Calcium sulphide ≥99% (metals basis excluding Mg) (max. 1% Mg)
Catalog # 43646.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Merck Index:14,1707
Melting Pt: Dec.
Density:2.5
ADR:4.3,III
:72.14 g/mol
EINECS:243-873-5
UN:3134
:GHS02
:GHS07
Storage Temperature:Ambient
CAS Number:20548-54-3
Formula:CaS
MDL Number:MFCD00015990