

Calcium hydroxide, Supelco®
Catalog # 1.02111.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Solubility:1.7 g/l (20 °C)
- Melting Point:550 °C (decomposition)
- Molar Mass:74.09 g/mol
- Bulk Density:400 kg/m3
- Boiling Point:2850 °C (1013 hPa) (decomposition)
- Vapor Pressure:(20 °C)
- Density:2.24 g/cm3 (20 °C)
- pH:12.6 (H2O, 20 °C) (saturated solution)
Specification Test Results
About this item
:
:74.09 g/mol
Merck Index:13,01675
Melting Pt:550 °C
Density:2.24 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-137-3
UN:3262
ADR:8,III
:M-Clarity™ quality level = MQ100
:GHS05
:GHS07
CAS Number:1305-62-0
Formula:Ca(OH)₂
MDL Number:MFCD00010901