Bestelling invoeren
Netherlands
ContactUsLinkComponent
Magnesium hydroxide carbonate light
Magnesium hydroxide carbonate light
Catalog # ROTH3530.1
Supplier:  Roth Carl
CAS Number:  
Magnesium hydroxide carbonate light
Catalog # ROTH3530.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Verpakking:
    Glass bottle for solids
  • VPE:
    2,5 kg
  • Seq:
    4076

Specifications

About this item

 :
 :84.31 g/mol
Density:2,16 g/cm³ (20 °C)
Storage Temperature:Kamertemperatuur
EINECS:235-192-7
CAS Number:12125-28-9
 :MFCD00149783
Formula:MgCO₃ (linear)