
Calcium hexaboride ≥99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12007-99-7
:
Formula:B₆Ca
MDL Number:MFCD00148904
:104.94 g/mol
Melting Pt:2235 °C
Density:2.30
EINECS:234-525-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together